This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PHF20 SIRNA Assays ›
Gene Symbol:
Phf20
Gene Name:
PHD finger protein 20
Gene Aliases:
6820402O20Rik
Chromosome Location:
Chr.2: 156196647 - 156309953 on Build GRCm38
UniGene ID:
Mm.427078
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s106020

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Phf20
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172674.2 NP_766262.2 3 313
GenBank mRNA AK033017.1 - 2 112
AK038573.1 - 3 220
AK045309.1 - 3 232
AK142424.1 - 3 205
BC060121.1 - 3 193 6413021
CJ176261.1 - 3 314

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
DNA binding
zinc ion binding
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
metal ion binding
histone acetyltransferase activity (H4-K16 specific)

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline