This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NDUFB6 SIRNA Assays ›
Gene Symbol:
Ndufb6
Gene Name:
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6
Gene Aliases:
Gm137
Chromosome Location:
Chr.4: 40270663 - 40279368 on Build GRCm38
UniGene ID:
Mm.1103
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s106379

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ndufb6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033305.2 NP_001028477.1 1, 2 209
GenBank mRNA AK146762.1 - 1, 2 211
BC147062.1 - 1, 2 198 9056053
BC147063.1 - 1, 2 198 9056059

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s)

transport
oxidation-reduction process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-2ck89:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline