siRNA Details
| Gene Symbol: Cyp2j13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145548.4 | NP_663523.2 | 2 | 302 | ||
| GenBank mRNA | AK143903.1 | - | 2 | 303 | ||
| AK143981.1 | - | 2 | 299 | |||
| AK162358.1 | - | 306 | ||||
| AK165405.1 | - | 2 | 302 | |||
| AK165469.1 | - | 2 | 280 | |||
| AK165473.1 | - | 2 | 280 | |||
| BC016446.1 | - | 2 | 292 | 4235131 | ||

Cyp2j13
Genomic Map