This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CYP2J13 SIRNA Assays ›
Gene Symbol:
Cyp2j13
Gene Name:
cytochrome P450, family 2, subfamily j, polypeptide 13
Gene Aliases:
-
Chromosome Location:
Chr.4: 96042660 - 96077566 on Build GRCm38
UniGene ID:
Mm.268175
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s106461

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyp2j13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145548.4 NP_663523.2 8 1307
GenBank mRNA AK143903.1 - 8 1308
AK143981.1 - 8 1304
AK162358.1 - 1311
AK165405.1 - 8 1307
AK165469.1 - 8 1285
AK165473.1 - 8 1285

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

xenobiotic metabolic process
biological_process
arachidonic acid metabolic process
exogenous drug catabolic process
oxidation-reduction process
molecular_function
arachidonic acid epoxygenase activity
steroid hydroxylase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
oxygen binding
heme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-4xrvm:80/100.66.79.185:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0