siRNA Details
| Gene Symbol: Wdtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199306.4 | NP_955010.1 | 13 | 1831 | ||
| GenBank mRNA | AK131700.1 | - | 15 | 1987 | ||
| AK172431.1 | - | 13 | 1877 | |||
| AK177370.1 | - | 2 | 204 | |||
| AK201097.1 | - | 1 | 95 | |||
| BC034833.1 | - | 6 | 754 | 5099225 | ||
| BC048824.1 | - | 13 | 1625 | 4188979 | ||
| BC057107.1 | - | 12 | 1474 | 5697877 | ||

Wdtc1
Genomic Map