siRNA Details
| Gene Symbol: Wdtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199306.4 | NP_955010.1 | 14 | 2034 | ||
| GenBank mRNA | AK131700.1 | - | 16 | 2190 | ||
| AK172431.1 | - | 14 | 2080 | |||
| BC034833.1 | - | 7 | 957 | 5099225 | ||
| BC048824.1 | - | 14 | 1828 | 4188979 | ||
| BC057107.1 | - | 13 | 1677 | 5697877 | ||

Wdtc1
Genomic Map