siRNA Details
| Gene Symbol: Haus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146159.1 | NP_666271.1 | 3 | 1260 | ||
| GenBank mRNA | AK049414.1 | - | 2 | 2230 | ||
| AK161756.1 | - | 2 | 287 | |||
| BC023882.1 | - | 3 | 1292 | 5354698 | ||
| BC025497.1 | - | 3 | 1278 | 5320270 | ||
| BC027393.1 | - | 3 | 1260 | 4948448 | ||

Haus3
Genomic Map