This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CYTL1 SIRNA Assays ›
Gene Symbol:
Cytl1
Gene Name:
cytokine-like 1
Gene Aliases:
4930443F05Rik, 4Cytl1, C17, Cyt1, Gm147
Chromosome Location:
Chr.5: 37735519 - 37739820 on Build GRCm38
UniGene ID:
Mm.76407
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s106780

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cytl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081106.1 NP_001074575.1 2 225
GenBank mRNA AK076770.1 - 1 117
BC063103.1 - 2 185
BC116224.1 - 2 199 40091368
BC116225.1 - 2 199 40091369
EF108311.1 - 2 196

Back To Top

More Information


Panther Classification:

Molecular Function -

cytokine intercellular signal molecule

Gene Ontology Categories:

Function(s)

chondrocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
inner ear development
chondroitin sulfate proteoglycan biosynthetic process
positive regulation of sequence-specific DNA binding transcription factor activity
cartilage homeostasis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-vzlfm:80/100.66.78.148:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0