siRNA Details
| Gene Symbol: BC037034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153161.3 | NP_694801.2 | 8 | 1394 | ||
| GenBank mRNA | AK046882.1 | - | 5 | 1854 | ||
| AK138960.1 | - | 8 | 1338 | |||
| BC037034.1 | - | 7 | 797 | 4505093 | ||
| BC040828.1 | - | 1 | 558 | |||
| BC044860.1 | - | 1 | 570 | |||

BC037034
Genomic Map