siRNA Details
| Gene Symbol: Zfp809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164624.1 | NP_001158096.1 | 5 | 720 | ||
| NM_172763.3 | NP_766351.3 | 5 | 729 | |||
| GenBank mRNA | AK044107.1 | - | 3 | 516 | ||
| AK049344.1 | - | 5 | 681 | |||
| AK089532.1 | - | 5 | 884 | |||
| BC099418.1 | - | 5 | 661 | 30525533 | ||
| CK389669.1 | - | 2 | 361 | |||

Zfp809
Genomic Map