siRNA Details
| Gene Symbol: Slc17a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276452.1 | NP_001263381.1 | 11 | 1390 | ||
| NM_172773.3 | NP_766361.1 | 11 | 1468 | |||
| GenBank mRNA | AK014522.1 | - | 1 | 926 | ||
| AK029102.1 | - | 11 | 1451 | |||
| AK169868.1 | - | 11 | 1417 | |||
| BC058785.1 | - | 11 | 1360 | 6309785 | ||

Slc17a5
Genomic Map