siRNA Details
| Gene Symbol: Igkv4-90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X14098.1 | - | 314 | |||
| Gene Symbol: Igkv4-57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | DQ372765.1 | - | 329 | |||
| DQ372805.1 | - | 329 | ||||

Igkv4-90
Genomic Map