siRNA Details
| Gene Symbol: Ccdc55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012309.2 | NP_001012309.1 | 6 | 588 | ||
| GenBank mRNA | AK046155.1 | - | 6 | 574 | ||
| AK078762.1 | - | 6 | 574 | |||
| AK078945.1 | - | 6 | 575 | |||
| AK168573.1 | - | 6 | 574 | |||
| BC016601.1 | - | 6 | 563 | |||
| BC089560.1 | - | 6 | 621 | 30300075 | ||

Ccdc55
Genomic Map