This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PDE4D SIRNA Assays ›
Gene Symbol:
Pde4d
Gene Name:
phosphodiesterase 4D, cAMP specific
Gene Aliases:
9630011N22Rik, Dpde3
Chromosome Location:
Chr.13: 108654177 - 109955969 on Build GRCm38
UniGene ID:
Mm.434429
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s108874

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde4d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011056.3 NP_035186.1 7 782
GenBank mRNA AF038896.1 - 5 464
AF536978.1 - 7 782
AK039354.1 - 5 376
AK039692.1 - 5 376
AK081013.1 - 6 1083
AY388962.1 - 5 395
BC132107.1 - 6 650 40130384
BC132109.1 - 6 650 40130386
EU489880.1 - 5 638

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001197220, NM_001104631, NM_001197219, NM_001197218, NM_001165899, and others.
This siRNA also targets the following transcripts in Rat: NM_001113328, NM_001113329, NM_001113334, NM_017032, NM_001113332, and others.

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

regulation of heart rate
cAMP catabolic process
smooth muscle contraction
signal transduction
aging
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
neutrophil chemotaxis
negative regulation of peptidyl-serine phosphorylation
multicellular organism growth
negative regulation of heart contraction
leukocyte migration
regulation of ryanodine-sensitive calcium-release channel activity
cellular response to lipopolysaccharide
cellular response to epinephrine stimulus
regulation of cardiac muscle cell contraction
regulation of cell communication by electrical coupling involved in cardiac conduction
negative regulation of relaxation of cardiac muscle
3',5'-cyclic-nucleotide phosphodiesterase activity
protein binding
phosphoric diester hydrolase activity
hydrolase activity
ion channel binding
metal ion binding
ATPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jspp5:80/100.66.79.6:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0