siRNA Details
| Gene Symbol: Ccdc112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160399.1 | NP_001153871.1 | 3 | 413 | ||
| GenBank mRNA | CJ174591.1 | - | 3 | 390 | ||
| DT929545.1 | - | 3 | 212 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc112| Gene Symbol: Ccdc112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160399.1 | NP_001153871.1 | 3 | 413 | ||
| GenBank mRNA | CJ174591.1 | - | 3 | 390 | ||
| DT929545.1 | - | 3 | 212 | |||