siRNA Details
| Gene Symbol: Slc16a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172838.3 | NP_766426.1 | 2, 3 | 299 | ||
| GenBank mRNA | AK028284.1 | - | 2, 3 | 297 | ||
| AK029076.1 | - | 2, 3 | 299 | |||
| BC111902.1 | - | 1, 2 | 252 | 40062303 | ||
| BC113803.1 | - | 1, 2 | 252 | 40062304 | ||

Slc16a12
Genomic Map