siRNA Details
| Gene Symbol: Kcnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081027.2 | NP_001074496.2 | 11 | 1028 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnt2| Gene Symbol: Kcnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081027.2 | NP_001074496.2 | 11 | 1028 | ||