This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DZANK1 SIRNA Assays ›
Gene Symbol:
Dzank1
Gene Name:
double zinc ribbon and ankyrin repeat domains 1
Gene Aliases:
2810039F03Rik, 6330439K17Rik, 6430408C01, AI851115, Ankrd64
Chromosome Location:
Chr.2: 144470555 - 144527425 on Build GRCm38
UniGene ID:
Mm.442156
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s109601

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dzank1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172859.2 NP_766447.2 10 1059
GenBank mRNA AK032182.1 - 10 1059
AK032632.1 - 10 1050
AK163994.1 - 10 1050
BC141156.1 - 10 958 9055768
BC145464.1 - 10 958 9054026

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy in response to mitochondrial depolarization
molecular_function
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline