siRNA Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081338.1 | NP_001074807.1 | 17 | 1946 | ||
| GenBank mRNA | AK049859.1 | - | 17 | 2001 | ||
| AK220350.1 | - | 15 | 1694 | |||
| BC116639.1 | - | 16 | 1905 | 40061282 | ||

L3mbtl1
Genomic Map