This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR1200 SIRNA Assays ›
Gene Symbol:
Olfr1200 Olfr1196 Olfr1197
Gene Name:
olfactory receptor 1200
olfactory receptor 1196
olfactory receptor 1197
Gene Aliases:
MOR225-12
MOR225-5
MOR225-10P, MOR225-14
Chromosome Location:
Chr.2: 88767303 - 88768313 on Build GRCm38
Chromosome Location:
Chr.2: 88700383 - 88701327 on Build GRCm38
Chromosome Location:
Chr.2: 88728653 - 88729597 on Build GRCm38
UniGene ID:
Mm.377285
Mm.377510
Mm.377954
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s110995

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 404.00
Your Price
Online offer: 391.65
391.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1200
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005227.2 NP_001005227.2 1 415
Gene Symbol: Olfr1196
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146464.2 NP_666675.2 1 415
Gene Symbol: Olfr1197
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005225.1 NP_001005225.1 1 415

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000344, NM_001000636, NM_001000638, XM_006234496.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline