This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR748 SIRNA Assays ›
Gene Symbol:
Olfr748 Olfr746
Gene Name:
olfactory receptor 748
olfactory receptor 746
Gene Aliases:
MOR106-9P
MOR106-12
Chromosome Location:
Chr.14: 50710332 - 50711255 on Build GRCm38
Chromosome Location:
Chr.14: 50653239 - 50654183 on Build GRCm38
UniGene ID:
Mm.377352
Mm.377417
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s111279

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr748
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011837.2 NP_001011837.2 1 596
Gene Symbol: Olfr746
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146298.2 NP_666410.2 1 596

Back To Top

More Information


Set Membership:

G-protein coupled receptor

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
cell-cell signaling
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
adenylate cyclase-activating adrenergic receptor signaling pathway
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline