This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR125 SIRNA Assays ›
Gene Symbol:
Olfr125
Gene Name:
olfactory receptor 125
Gene Aliases:
MOR218-8
Chromosome Location:
Chr.17: 37834911 - 37836028 on Build GRCm38
UniGene ID:
Mm.377416
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s111551

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr125
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146290.2 NP_666402.2 1 598
GenBank mRNA BC116810.1 - 1 598 40126129
BC116812.1 - 1 598 40126131

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000557.

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline