This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MAP2K3 SIRNA Assays ›
Gene Symbol:
MAP2K3
Gene Name:
mitogen-activated protein kinase kinase 3
Gene Aliases:
MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
Chromosome Location:
Chr.17: 21284656 - 21315240 on Build GRCh38
UniGene ID:
Hs.514012
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s11175
Validated

Purification
Size
Availability Inventoried
Catalog # 4390824
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAP2K3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316332.1 NP_001303261.1 4 535
NM_002756.4 NP_002747.2 3 152
NM_145109.2 NP_659731.1 3 410
XM_005256723.2 XP_005256780.1 4 450
XM_011523958.2 XP_011522260.1 3 209
XM_011523959.1 XP_011522261.1 5 417
XM_017024857.1 XP_016880346.1 3 279
XM_017024858.1 XP_016880347.1 4 267
XM_017024859.1 XP_016880348.1 5 607
GenBank mRNA AB451226.1 - 3 145
AB451349.1 - 3 145
AK093838.1 - 4 454
AK223490.1 - 3 285
AK315505.1 - 3 355
BC032478.1 - 3 345 5215093
BM836867.1 - 3 390
CR536514.1 - 3 145
CR541793.1 - 3 145
D87116.1 - 3 202
DA131175.1 - 3 410
DA987047.1 - 3 153
GQ225578.1 - 3 145
L36719.1 - 5 395
U66839.1 - 3 271

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00177127_m1

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
activation of MAPK activity
inflammatory response
signal transduction
peptidyl-tyrosine phosphorylation
proteolysis in other organism
regulation of cytokine biosynthetic process
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
cardiac muscle contraction
receptor signaling protein serine/threonine kinase activity
MAP kinase kinase activity
protein tyrosine kinase activity
protein binding
ATP binding
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline