siRNA Details
Gene Symbol: Olfr945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146506.1 | NP_666717.1 | 1 | 801 | ||
GenBank mRNA | BC128004.1 | - | 1 | 805 | 40135946 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146506.1 | NP_666717.1 | 1 | 801 | ||
GenBank mRNA | BC128004.1 | - | 1 | 805 | 40135946 |