This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR972 SIRNA Assays ›
Gene Symbol:
Olfr972
Gene Name:
olfactory receptor 972
Gene Aliases:
MOR171-17
Chromosome Location:
Chr.9: 39873277 - 39874221 on Build GRCm38
UniGene ID:
Mm.271456
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s112414

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr972
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146610.1 NP_666821.1 1 436

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
odorant binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-jrfm9:80/100.66.76.154:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0