This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR403 SIRNA Assays ›
Gene Symbol:
Olfr403 Olfr43
Gene Name:
olfactory receptor 403
olfactory receptor 43
Gene Aliases:
IA7, MOR125-5_p, Olfr43
IA7, MOR125-1, MOR125-5_p, Olfr403
Chromosome Location:
Chr.11: 74186249 - 74196446 on Build GRCm38
Chromosome Location:
Chr.11: 74206198 - 74207289 on Build GRCm38
UniGene ID:
Mm.246537
Mm.453816
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s112707

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr403
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207622.1 NP_997505.1 1 448
GenBank mRNA AF102517.1 - 1 259
BC104289.1 - 1 448 40042769
BC104290.1 - 1 448 40042770
Gene Symbol: Olfr43
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146711.2 NP_666922.2 1 523
GenBank mRNA BC125464.1 - 1 523 40129979
BC125468.1 - 1 523 40129983

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-lqf76:80/100.66.77.18:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline