siRNA Details
| Gene Symbol: Olfr905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146804.2 | NP_667015.2 | 1 | 137 | ||
| GenBank mRNA | BC150476.1 | - | 1 | 141 | 40134740 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr905| Gene Symbol: Olfr905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146804.2 | NP_667015.2 | 1 | 137 | ||
| GenBank mRNA | BC150476.1 | - | 1 | 141 | 40134740 | |