siRNA Details
| Gene Symbol: Olfr658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147049.4 | NP_667260.1 | 2 | 1960 | ||
| GenBank mRNA | AK033992.1 | - | 2 | 1961 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr658| Gene Symbol: Olfr658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147049.4 | NP_667260.1 | 2 | 1960 | ||
| GenBank mRNA | AK033992.1 | - | 2 | 1961 | ||