siRNA Details
| Gene Symbol: Olfr585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147087.2 | NP_667298.2 | 1 | 601 | ||
| GenBank mRNA | BC145897.1 | - | 1 | 627 | 40131200 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr585| Gene Symbol: Olfr585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147087.2 | NP_667298.2 | 1 | 601 | ||
| GenBank mRNA | BC145897.1 | - | 1 | 627 | 40131200 | |
Set Membership: |
|