This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OLFR427 SIRNA Assays ›
Gene Symbol:
Olfr427 Olfr426
Gene Name:
olfactory receptor 427
olfactory receptor 426
Gene Aliases:
MOR105-7, MOR105-9
GA_x6K02T2P20D-21063569-21062619, MOR105-13_p, MOR105-8
Chromosome Location:
Chr.1: 174099460 - 174100407 on Build GRCm38
Chromosome Location:
-
UniGene ID:
Mm.377354
Mm.489574
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID s113845

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr427
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207158.1 NP_997041.1 1 483
GenBank mRNA BC104104.2 - 1 483 40043137
BC106808.2 - 1 483 40042917
Gene Symbol: Olfr426
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206926.1 NP_001193855.1 1 483

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-brk9k:80/100.66.78.44:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0