siRNA Details
| Gene Symbol: Olfr427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207158.1 | NP_997041.1 | 1 | 343 | ||
| GenBank mRNA | BC104103.2 | - | 1 | 343 | 40043136 | |
| BC104104.2 | - | 1 | 343 | 40043137 | ||
| BC106807.2 | - | 1 | 343 | 40042912 | ||
| BC106808.2 | - | 1 | 343 | 40042917 | ||

Olfr427
Genomic Map