This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LEAP2 SIRNA Assays ›
Gene Symbol:
Leap2
Gene Name:
liver-expressed antimicrobial peptide 2
Gene Aliases:
leap-2
Chromosome Location:
Chr.11: 53422181 - 53423136 on Build GRCm38
UniGene ID:
Mm.24283
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s113863

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 5,354.00
Your Price
Online offer:
5354.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Leap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153069.3 NP_694709.1 2 208
GenBank mRNA AJ409055.1 - 2 208
BC089593.1 - 2 201 6749700
W82161.1 - 2 242

Back To Top

More Information


Gene Ontology Categories:

Function(s)

defense response to bacterium

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0