This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM107A SIRNA Assays ›
Gene Symbol:
Fam107a
Gene Name:
family with sequence similarity 107, member A
Gene Aliases:
DRR1
Chromosome Location:
Chr.14: 8296270 - 8318018 on Build GRCm38
UniGene ID:
Mm.256058
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s114077

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam107a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183187.3 NP_899010.1 2 400
GenBank mRNA AK044219.1 - 2 170
AK165722.1 - 3 587
BC055107.1 - 2 400 6532624

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline