siRNA Details
| Gene Symbol: Mroh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033365.2 | NP_001028537.1 | 7 | 701 | ||
| GenBank mRNA | AK158799.1 | - | 7 | 702 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mroh5| Gene Symbol: Mroh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033365.2 | NP_001028537.1 | 7 | 701 | ||
| GenBank mRNA | AK158799.1 | - | 7 | 702 | ||