siRNA Details
| Gene Symbol: Abat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170978.1 | NP_001164449.1 | 2, 3 | 259 | ||
| NM_172961.3 | NP_766549.2 | 2, 3 | 259 | |||
| GenBank mRNA | AK036128.1 | - | 2, 3 | 204 | ||
| AK083062.1 | - | 2, 3 | 206 | |||
| AK132282.1 | - | 2, 3 | 208 | |||
| AK160802.1 | - | 2, 3 | 207 | |||
| AK165348.1 | - | 2, 3 | 209 | |||
| BC058079.1 | - | 2, 3 | 199 | 6826741 | ||
| BC058521.1 | - | 2, 3 | 199 | 6825487 | ||
| CX237932.1 | - | 2, 3 | 259 | |||

Abat
Genomic Map