This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GCDH SIRNA Assays ›
Gene Symbol:
Gcdh
Gene Name:
glutaryl-Coenzyme A dehydrogenase
Gene Aliases:
9030411L18, AI266902, D17825, GCD
Chromosome Location:
Chr.8: 84886387 - 84893921 on Build GRCm38
UniGene ID:
Mm.2475
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s114508

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gcdh
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044744.1 NP_001038209.2 11 1423
NM_008097.2 NP_032123.3 10 1480
GenBank mRNA AK078866.1 - 8 1033
AK079500.1 - 4 531
AK165406.1 - 10 1306
BC061158.1 - 10 1310 30314273
U18992.1 - 10 1261

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

acyl-CoA metabolic process
metabolic process
fatty acid oxidation
fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
glutaryl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
flavin adenine dinucleotide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nzkhn:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline