This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other A230046K03RIK SIRNA Assays ›
Gene Symbol:
A230046K03Rik
Gene Name:
RIKEN cDNA A230046K03 gene
Gene Aliases:
AA589518, Kiaa1033, mKIAA1033
Chromosome Location:
Chr.10: 83543941 - 83596486 on Build GRCm38
UniGene ID:
Mm.491194
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115123

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: A230046K03Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033375.2 NP_001028547.2 27 2879
GenBank mRNA AK038582.1 - 27 2837
AK043911.1 - 27 2850
AK129269.1 - 22 2409
AK145000.1 - 27 2861
AK161247.1 - 4 327
AK166233.1 - 27 2852
BC020314.2 - 24 2514
BC026467.1 - 16 1840
BC145215.1 - 27 3074

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
endosome organization
protein transport
endosomal transport
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline