siRNA Details
| Gene Symbol: Fndc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207636.2 | NP_997519.2 | 8 | 1202 | ||
| GenBank mRNA | AK032733.1 | - | 8 | 1174 | ||
| AK048586.1 | - | 8 | 1205 | |||
| AK089380.1 | - | 8 | 1167 | |||
| AK154790.1 | - | 8 | 971 | |||
| AK172458.1 | - | 8 | 1066 | |||
| BC144880.1 | - | 8 | 1010 | |||
| DQ192036.1 | - | 8 | 1203 | |||

Fndc3a
Genomic Map