This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FSD1L SIRNA Assays ›
Gene Symbol:
Fsd1l
Gene Name:
fibronectin type III and SPRY domain containing 1-like
Gene Aliases:
A230072O16Rik, Ccdc10, Csdufd1, Fsd1nl, OTTMUSG00000007002
Chromosome Location:
Chr.4: 53629929 - 53707009 on Build GRCm38
UniGene ID:
Mm.103464
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115292

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 296.00
296.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fsd1l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195284.1 NP_001182213.1 4 436
NM_176966.4 NP_795940.1 4 436
GenBank mRNA AK038893.1 - 4 414
AK052914.1 - 4 479
AK140020.1 - 4 420
BB662298.1 - 4 474
BC130265.1 - 4 352 40142990

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_207647, NM_031919, NM_001145313, BP219536, BP365251, and others.

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gtpqw:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline