This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other USP37 SIRNA Assays ›
Gene Symbol:
Usp37
Gene Name:
ubiquitin specific peptidase 37
Gene Aliases:
4932415L06Rik, C330008N13Rik
Chromosome Location:
Chr.1: 74435510 - 74544286 on Build GRCm38
UniGene ID:
Mm.471601
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115300

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 404.00
Your Price
Online offer: 391.65
391.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Usp37
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176972.4 NP_795946.2 14 1798
GenBank mRNA AK030013.1 - 14 1799
AK035640.1 - 13 1782
AK173210.1 - 10 1274
BC114208.1 - 13 1762
BC139091.1 - 14 1740 8862113

Back To Top

More Information


Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
proteolysis
ubiquitin-dependent protein catabolic process
cell cycle
mitotic nuclear division
protein deubiquitination
protein K11-linked deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
cell division
regulation of proteasomal protein catabolic process
protein K48-linked deubiquitination
cysteine-type endopeptidase activity
thiol-dependent ubiquitin-specific protease activity
peptidase activity
cysteine-type peptidase activity
hydrolase activity
protein kinase binding
thiol-dependent ubiquitinyl hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0