This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC35E2 SIRNA Assays ›
Gene Symbol:
Slc35e2
Gene Name:
solute carrier family 35, member E2
Gene Aliases:
A530082C11Rik, AI957035
Chromosome Location:
Chr.4: 155600592 - 155623340 on Build GRCm38
UniGene ID:
Mm.306018
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115770

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc35e2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177186.4 NP_796160.1 5 1172
GenBank mRNA AK048716.1 - 5 1041
AK080215.1 - 5 1191
AK157354.1 - 5 1068
BC058728.1 - 5 1172 6407259

Back To Top

More Information


Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

biological_process
anion transmembrane transport
molecular_function
antiporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0