This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ISLR2 SIRNA Assays ›
Gene Symbol:
Islr2
Gene Name:
immunoglobulin superfamily containing leucine-rich repeat 2
Gene Aliases:
B930052A04Rik, Linx, Mbu-3, mKIAA1465
Chromosome Location:
Chr.9: 58196297 - 58204319 on Build GRCm38
UniGene ID:
Mm.186499
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115788

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Islr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161535.1 NP_001155007.1 2 2322
NM_001161536.1 NP_001155008.1 4 2503
NM_001161537.1 NP_001155009.1 3 2494
NM_001161538.1 NP_001155010.1 4 2271
NM_001161539.1 NP_001155011.1 4 2233
NM_001161540.1 NP_001155012.1 3 2189
NM_001161541.1 NP_001155013.1 3 2080
NM_177193.5 NP_796167.2 3 2313
GenBank mRNA AK043849.1 - 1 147
AK047348.1 - 1 1801
AK129367.1 - 3 1999
BC059068.1 - 3 2166 6825665
BC096531.1 - 2 2322 6826287
EU497918.1 - 3 2130

Back To Top

More Information


Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
positive regulation of axon extension
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0