This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other A330050F15RIK SIRNA Assays ›
Gene Symbol:
A330050F15Rik
Gene Name:
RIKEN cDNA A330050F15 gene
Gene Aliases:
-
Chromosome Location:
Chr.17: 69437591 - 69489233 on Build GRCm38
UniGene ID:
Mm.400254
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s115898

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: A330050F15Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145192.1 NP_001138664.1 2 522
GenBank mRNA AK031811.1 - 2 631
AK039494.1 - 2 465

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein localization
protein kinase A binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline