This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VPRBP SIRNA Assays ›
Gene Symbol:
Vprbp
Gene Name:
Vpr (HIV-1) binding protein
Gene Aliases:
AI447437, B930007L02Rik, mKIAA0800
Chromosome Location:
Chr.9: 106814172 - 106880992 on Build GRCm38
UniGene ID:
Mm.440089
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s116041

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vprbp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001015507.2 NP_001015507.1 7 884
GenBank mRNA AK122372.1 - 7 847
BC065119.1 - 7 884 6853519

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191798.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
viral process
phosphorylation
chromatin modification
B cell differentiation
V(D)J recombination
cell competition in a multicellular organism
histone H2A-T120 phosphorylation
nucleotide binding
protein serine/threonine kinase activity
ATP binding
kinase activity
transferase activity
histone kinase activity (H2A-T120 specific)

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-pcbtx:80/100.66.76.251:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0