This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM26 SIRNA Assays ›
Gene Symbol:
Tmem26
Gene Name:
transmembrane protein 26
Gene Aliases:
3230401A06, AI195372
Chromosome Location:
Chr.10: 68723746 - 68782655 on Build GRCm38
UniGene ID:
Mm.2479
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s116080

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem26
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177794.3 NP_808462.2 4 610
GenBank mRNA AK028283.1 - 4 611
AK084351.1 - 4 588
AK143866.1 - 4 588
BC117502.1 - 5 614 40106131
BC117503.1 - 4 532 40106128

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107623.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-zcg68:80/100.66.77.208:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline