siRNA Details
| Gene Symbol: Dnah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081330.1 | NP_001074799.1 | 65 | 10187 | ||
| GenBank mRNA | AK029791.1 | - | 12 | 1660 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnah2| Gene Symbol: Dnah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081330.1 | NP_001074799.1 | 65 | 10187 | ||
| GenBank mRNA | AK029791.1 | - | 12 | 1660 | ||