This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FREM1 SIRNA Assays ›
Gene Symbol:
Frem1
Gene Name:
Fras1 related extracellular matrix protein 1
Gene Aliases:
BC037594, D430009N09, D630008K06, crf11, eye<m02Jus>, eyes2, heb, qbrick
Chromosome Location:
Chr.4: 82897920 - 83052548 on Build GRCm38
UniGene ID:
Mm.242337
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s116502

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 400.00
Your Price
Online offer: 388.65
388.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Frem1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198811.1 NP_001185740.1 23 4595
NM_177863.4 NP_808531.2 22 4385
GenBank mRNA AB160986.1 - 23 4595
AJ616838.1 - 22 4385
AK052629.1 - 6 846
BC138427.1 - 22 4296 8861447
BC145274.1 - 22 4296

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell communication
cell adhesion
cell-matrix adhesion
multicellular organism development
craniofacial suture morphogenesis
carbohydrate binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-wzvtq:80/100.66.78.229:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline