siRNA Details
| Gene Symbol: B4galnt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177897.3 | NP_808565.2 | 16 | 2550 | ||
| GenBank mRNA | AB114827.1 | - | 16 | 2550 | ||
| BC038881.1 | - | 2 | 497 | |||
| BC062653.1 | - | 12 | 2240 | |||
| BC079533.1 | - | 5 | 1453 | |||

B4galnt4
Genomic Map