siRNA Details
| Gene Symbol: Susd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101510.1 | NP_001094980.1 | 3 | 525 | ||
| GenBank mRNA | AK132299.1 | - | 3 | 446 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Susd5| Gene Symbol: Susd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101510.1 | NP_001094980.1 | 3 | 525 | ||
| GenBank mRNA | AK132299.1 | - | 3 | 446 | ||