This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KCTD16 SIRNA Assays ›
Gene Symbol:
Kctd16
Gene Name:
potassium channel tetramerisation domain containing 16
Gene Aliases:
4930434H12Rik, Gm1267
Chromosome Location:
Chr.18: 40258361 - 40531184 on Build GRCm38
UniGene ID:
Mm.76366
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s118253

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kctd16
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026135.1 NP_080411.1 2 986
GenBank mRNA AK005863.1 - 1 38
AK006368.1 - 1 38
AK015313.1 - 2 320
AK220381.1 - 2 960
BC049734.1 - 3 254

Back To Top

More Information


Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of G-protein coupled receptor protein signaling pathway
protein homooligomerization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline